Mortar and Pestle Sets
WARD470019-978 12091-252
470019-978EA 469.83 MXN
470019-978 470148-960
Mortar and Pestle Sets
Morteros y manos de mortero Morteros con manos de mortero
Oversized Pestle Provides a Comfortable Grip when Grinding
  • Mortar with lip is glazed outside
  • Pestle is glazed except for the grinding surface
  • Choose from two sizes
  • Autoclavable