Slotted Masses Set of 12, Stainless Steel
Proveedor: VWR International
PH0037HSS
470014-354EA 1395.27 MXN
470014-354
Slotted Masses Set of 12, Stainless Steel
Pesas Pesas de calibración
Twelve of the most widely used weights on a rack for convenient storage and transport.
  • Includes 12 individually calibrated slotted masses totaling 1110 g
  • Made from high-grade stainless steel
  • Mass values are printed on each piece for easy identification
  • Includes plastic and metal rack with handle for convenient storage and transport
This stainless steel slotted mass set is placed on a plastic and metal rack with handle. Set of 12 masses includes 1 g (1), 2 g (2), 5 g (1), 10 g (1), 20 g (2), 50 g (1) 100 g (1), 200 g (2), 500 g (1). Total mass: 1110 g.