Spirogyra, Scalariform Conjugation Slide
470180-986
: Avantor
- Description:Spirogyra, Scalariform Conjugation Slide
- Cat. no.:470180-986
- Stain:Fast Green and Mallory's
- Supplier No.:WARD470180-986
- Orientation:Whole mount
Illustrates both forms of reproduction
- Identifying features clearly distinguishable
- Rigorous quality control standards
Showing vegetative filaments, conjugation, and zygotes.