Order Entry
Spirogyra, Vegetative Slide
Spirogyra, Vegetative Slide
  470177-422
 :  Avantor
undefined
Spirogyra, Vegetative Slide
  470177-422
 :  Avantor
$116.28
  Each (1UN)
  • Description:
    Spirogyra, Vegetative Slide
  • Cat. no.:
    470177-422
  • Stain:
    Fast Green and Mallory's
  • Supplier No.:
    WARD470177-422
  • Orientation:
    Whole mount
Investigate spirogyra structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Cells with several chloroplasts; also showing nucleus, pyrenoids, and other cytoplasmic detail.