Standard Bunsen Burners
Proveedor: VWR International
C-1476/3 C-1484 C-1477
470148-792CS 4831.2 MXN
470148-792 470148-926 470148-654
Standard Bunsen Burners
Calentadores Mecheros de gas
These standard Bunsen burners are practical and economical.
  • Gas Inlet: 5/16" Diameter
  • Mixing Tube: ½" Diameter without Stabilizer
  • Mixing Tube: 7/10" Diameter with Stabilizer
  • Height: 6⅛" without Stabilizer
  • Height: 6⅜" with Stabilizer
They feature an aluminum mixing tube and a sturdy die-cast, nickel-plated base.
With a stabilizer, they have a height of 6⅛".
Neoprene Bunsen Burner Tubing is recommended.