Steel Rods
Proveedor: VWR International
WLS78306-20C WLS78306-20D S78306-20A WLS78306-20B
470123-468EA 169.3 MXN
470123-468 470123-466 470123-464 470123-462
Steel Rods
Varillas de retorta
Made of zinc plated steel; designed to fit T-Nesting Support Bases. One end threaded, with shoulder, to fit base.