The Effects of Drugs Kit
Proveedor: SCIENCE INTERACTIVE GROUP
2200
470008-010KT 2968.13 MXN
470008-010
The Effects of Drugs Kit
Pruebas de drogas
This kit enables students to observe a living experimental organism, describe the effects of seven drugs, determine a dose-response curve, and correlate test drugs to effects on humans.
  • Enables students to observe living organism
  • Develop Daphnia cultures
Students develop Daphnia cultures and expose them to nicotine, alcohol, caffeine, aspirin and three other drugs. From this data, students make dose-response graphs. Includes materials for 30 students and a teacher’s manual with 15 student worksheets. Note: Daphnia are required but not included.