H-B® DURAC® General Purpose Student Thermometers, Organic Liquid Fill
Proveedor: BEL-ART PRODUCTS SE
B62004-0700 B62002-0300
470006-496EA 129.12 MXN
470006-496 470105-090
H-B® DURAC® General Purpose Student Thermometers, Organic Liquid Fill
Termómetros Analogue Thermometers
Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries.
  • Safer than mercury thermometers
  • Contain red organic liquid against white back glass