SCIENCEWARE® Wide Mouth Wash Bottles
Proveedor: BEL-ART PRODUCTS SE
F11620-1000_SE F11620-0500_SE F11620-0250_SE F11620-0125_SE
470145-308CS 3014.64 MXN
470145-308 470145-310 470145-312 470145-314
SCIENCEWARE® Wide Mouth Wash Bottles
Frascos Frascos de lavado
Low-density polyethylene wash bottles have color-coded polypropylene closures for easy identification and wide mouths for easy filling.
  • Color-coded closures for quick identification
  • Wide-mouthed design
  • LDPE construction
Natural caps in 125, 250, 500, or 1000 ml sizes.