Wind-Up Metric Tape Measure
Proveedor: HORIZON SOLUTIONS CORP.
FM050CME
470106-460EA 1618.62 MXN
470106-460
Wind-Up Metric Tape Measure
Cintas métricas
Indestructible, High quality, Fiberglass Tape.
  • 14 mm wide tape
  • Graduated to 0.5 cm on one side
  • Other side has additional 2 mm graduations
Fiberglass tape contained in a plastic wind-up case with a recessed winding handle. Ideal for outdoor education because the fiberglass tape will not rust, kink or clog with dirt.