Flip Cap Aluminum Seals, WHEATON®
Proveedor: DWK Life Sciences
224208 224203 224202
16171-944CS 7295.13 MXN
16171-948 16171-944 16171-942
Flip Cap Aluminum Seals, WHEATON®
Tapas Tapones para viales de cromatografía
Flip cap seals provide tamper evidence
Flip cap tear-off seals remove completely allowing content to pour from bottle.
The red plastic top flips off easily with pressure from the thumb exposing the aluminum seal with center hole. Contents can be accessed by syringe.