SP Bel-Art H-B Easy-Read™ Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a part of SP
B60305-0100 B60304-0500 B60301-0200 B60303-0000 B60305-0000 B60304-0400 B60300-0000 B60301-0300 B60300-0100 B60301-0500 B60304-0300 B60301-0000 B60302-0000 B60301-0600 B60304-0000 B60301-0100 B60304-0200 B60302-0100 B60301-0400
89217-924EA 779.87 MXN
89217-916 89217-928 89217-918 89217-934 89217-912 89217-924 89217-914 89217-936 89217-926 89217-948 89217-930 89217-952 89217-920 89217-932 89217-910 89217-922 89217-950 CA10768-366 89082-219
SP Bel-Art H-B Easy-Read™ Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Termómetros Analogue Thermometers
EnviroKleen™ certified thermometers feature black, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, yellow back glass for easy readability and fewer reading errors.
  • Non-toxic and biodegradable liquid
  • Lead-free
  • Yellow back glass
  • Easy to read
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Información para pedidos: Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.