E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences
Proveedor: DWK Life Sciences
224728 224711 224746 224745 224700 224704 224708 W226060 224714 224751 224748 224730 224740 224731 224724 224712 224741 224734 224742 224702 224721 224732 224750 224722 224725 224726 224754 224752 224705 224744 224701 224706 224720 224710
66011-502PK 1548.04 MXN
66011-506 66011-528 66011-548 66011-504 66011-526 66011-546 66011-524 66011-502 66011-500 66011-544 66011-522 66011-520 66011-542 66011-564 66011-540 66011-562 66011-560 66011-518 66011-516 66011-538 66011-536 66011-558 66011-514 66011-535 66011-512 66011-556 66011-554 66011-532 66011-510 66011-552 66011-530 66011-550 83009-706 66011-508
E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences
Viales
Wheaton E-C Sample Vials are economically priced by offering vials without screw caps or with screw caps packaged separately.
  • Autoclavable vials and caps
Vials are offered with a choice of 14B rubber or PTFE faced 14B Rubber lined black phenolic caps. PTFE provides a totally inert inner seal and surface facing the sample or product.
Vials are offered in both clear and amber glass and are packaged in in shrink-wrapped packs of 100 vials to reduce contamination from corrugate.
Certificaciones: Clear vials conform to USP Type I and ASTM E438 Type 1, Class A requirements while amber vials conform to USP Type I requirements for light transmission to protect light sensitive samples.
Información para pedidos: Caps for vials without screw caps can be purchased separately.