VWR® Aerosol Filter Pipette Tips in Racks
Proveedor: VWR
VWR®
1156-965-306 1094-965-306
82003-192CS 16174.52 MXN
89000-008 82003-192
VWR® Aerosol Filter Pipette Tips in Racks
Puntas de pipetas
VWR signature aerosol filter pipette tips offer you a high-value option to protect your pipettors in all liquid handling applications.
  • Endotoxin-free filter
  • Porous filter design
  • Autoclavable
They include an endotoxin-free 10 µm patented porous filter. Certified to be free of Endotoxin, Oleamide, DNA and Nuclease (RNase/DNase). Tips are autoclavable.
Certificaciones: ISO 9001, CE IVD
Información para pedidos: 96 tips per rack, 10 racks per pack. 9600 tips per case.