Glass Fiber Prefilters, Corning
Proveedor: Corning
Corning®
431412 431410 431413 431411
97039-896CS 2727.51 MXN
97039-894 97039-896 97039-898 97039-900
Glass Fiber Prefilters, Corning
Filtros Filtros de fibras de vidrio
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.