Conductivity Standard, 10 µS/cm (10 µmho/cm) at 25 °C 4.7 ppm as NaCl, Ricca Chemical Company
Catalog # RC2236-20
Supplier: RICCA CHEMICAL CO
Small Business Enterprise
CAS Number:
Discontinued
Specifications
- Pack type:20 per package
- Size:Pack
- Cat. no.:RC2236-20
- Supplier No.:R2236000-20PK