- Volume:50 - 1000 µl
- Cat. no.:89233-098
- Increments:1 µl
- Accuracy:±1.5% at 100 µl, ±0.4% at 1000 µl
- Precision:±0.5% at 100 µL, ±0.15% at 1000 µL
- Supplier No.:735081
Biohit Picus is the smallest and lightest electronic pipette on the market, helping to avoid pipetting-caused symptoms like RSI (repetitive strain injury). The Picus handle design and location of operation buttons enable comfortable pipetting. The patented electronic tip ejection, one hand-operated Adjustment wheel, and the conveniently located Start button minimizes the force needed in pipetting. Repetitive and time-consuming pipetting can be done accurately and fast with serial dispensing modes and a new Tracker – a great help in usage such as microplate work. Due to the advanced DC-motor concept and built-in error contro,l Biohit Picus ensures the most accurate and precise pipetting results.
- Unique lightweight and compact design maximizes user convenience
- Conveniently located, one-touch operating-button
- Feather-light electronic tip ejection help minimize muscle strain
- Electronic piston control and brake ensure accurate, user-independent pipetting results
- Unique plate tracker increases reliability in microwell plate dispensing
- Comprehensive selection of pipetting modes offers a number of different liquid handling tasks quickly and easily
- Universal AC adaptor included (EU, UK, US/JPN, AUS, and CHN plugs)
Picus has been awarded the 2012 Red Dot Design Award.
Intuitive user interface provides a choice of five language options: English, French, German, Russian, and Chinese.
By guiding pipetting steps, the unique plate tracker increases reliability in correctly filling microwell plates. The pipet's repetitive blow-out function facilitates dispensing of the very last sample droplets.