Purifier® Horizontal Clean Benches, Labconco®
Proveedor: Labconco
3873023 3888408 3888432 3818508 3250011 3889721 3888423 3889722 3888420 3250008 3873002 3818532 3889701 3818506 3818501 3250031 3818522 3873026 3889705 3250020 3250029 3250025 3888409 3873022 3873021 3250006 3889706 3888428 3889708 3888400 3250005 3818523 3889725 3250003 3873008 3818505 3818531 3888406 3888422 3889709 3889731 3818528 3888411 3889723 3818512 3818520 3250032 3888421 3250026 3250001 3250023 3818526 3873009 3873032 3873029 3888403 3250012 3889700 3888429 3250021 3818521 3818525 3873012 3888401 3873000 3250028 3250022 3818511 3873001 3250009 3873005 5361150 3873028 3889720 3889729 3888412 3889703 3250000 3888405 3889732 3888425 3889711 3873020 3888431 3818509 3873003 3889728 3889726 3889712 3873031 3873025 3888426 3889702 3818529 3250002 3818502 3818500 3873011 3888402 3818503
89232-288EA 398255 MXN
89232-292 89232-290 89497-258 89497-256 89497-254 89497-252 89497-250 89232-298 89203-904 89232-296 89203-902 89203-900 89232-294 89203-888 89232-282 89203-886 89232-280 89497-248 89497-246 89497-244 89497-242 89497-240 89232-324 89232-322 89232-288 89232-320 89232-286 89232-284 89232-270 89232-318 89232-316 89232-314 97007-592 89232-312 89232-278 89232-310 89232-276 89232-274 89232-272 89232-260 89232-380 89497-260 89232-308 89232-306 89232-304 89232-302 89232-268 89232-388 89232-300 89232-266 89232-386 89232-264 89232-384 89232-262 89232-382 89232-258 89232-378 89232-256 89232-376 89232-254 89232-374 89232-252 89232-372 89232-250 89232-248 89232-368 89232-246 89232-366 89232-244 89232-364 89232-242 89232-362 89232-240 89232-360 89203-898 89203-896 89497-238 89203-894 89497-236 89203-892 89497-234 89203-890 89497-232 89497-230 89232-238 89232-358 89232-236 89232-356 89232-234 89232-354 89232-232 89232-352 89232-230 89232-350 89497-228 89497-226 89497-224 89497-222 89232-228 89232-226
Purifier® Horizontal Clean Benches, Labconco®
Vitrinas de gases y recintos Armarios de flujo laminar
The Purifier Horizontal Clean Bench protects your work
The angled profile design means the front panel is free of protrusions that could interfere with visibility or obstruct taller users. Puifier Horizontal Clean Benches feature double-gasket, negative pressure leak protection.
These clean benches include a 99.99% efficient HEPA, front-mounted light and blower switches and one year warranty on parts and labor. Replaceable prefilters trap large particles and extend filter life.
It is the ISO Class 5 work station of choice to protect materials that do not generate harmful aerosols or vapors, yet require a particulate-free environment. Purifier Horizontal Clean Benches are offered in 3', 4', 6' and 8' widths and are available with 21" and 26" interior depths. Factory-installed UV lamp with UV scren and interlocking safety switch are also available. Standard on all units is a Type 304 brushed stainless steel work surface and 1/4" thick safety glass side panels.
Certificaciones: ETL, CE listed.
Información para pedidos: Base stands are not included and need to be purchased separately. Height selection and adjustment must be completed before the enclosure is installed atop the stand.