Tuberculin Mini Glass Syringes, Cadence Science®
Proveedor: Cadence Science
5207
20068-036EA 1731.59 MXN
20068-036
Tuberculin Mini Glass Syringes, Cadence Science®
Jeringas Jeringas médicas
Tuberculin mini glass syringes are widely used in clinical and research procedures for the administration of precise low-volume doses ranging from 1/4mL to 2mL
Fabricated of chemically resistant borosilicate glass with easy-to-read accurate permanent calibration lines.
Información para pedidos: Syringes are individually boxed.