Order Entry
Accessories for Pipet-Aid® Pipette Controllers, Drummond
Accessories for Pipet-Aid® Pipette Controllers, Drummond
Certificates
Accessories for Pipet-Aid® Pipette Controllers, Drummond
Product Information
  • Catalog #
    Supplier #
    For Use With
    Supplier No.
    Availability
    Price
  • Replacement Filters, Nonsterile
    Replacement Filters, Nonsterile
    53498-205
    4-000-051
    Pipet-Aid
    4-000-051
    Log in for availability
    Log in for availability
    Paquete de 5
    Paquete de 5 - $935.83
    $935.83
    Paquete de 5
       $935.83
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Outer Shell for Tissue Culture Nosepiece Kit
    Outer Shell for Tissue Culture Nosepiece Kit
    53498-068
    4-000-053-A
    Pipet-Aid
    4-000-053-A
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,036.11
    $1,036.11
    Each (1UN)
       $1,036.11
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Tissue Culture Nosepiece Kit with 2 Filters
    Tissue Culture Nosepiece Kit with 2 Filters
    53498-065
    4-000-050
    Pipet-Aid
    4-000-050
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $2,692.65
    $2,692.65
    Each (1UN)
       $2,692.65
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Rechargeable NiMH Battery
    Rechargeable NiMH Battery
    53498-063
    4-000-035
    Pipet-Aid XP
    4-000-035
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $996.52
    $996.52
    Each (1UN)
       $996.52
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Replacement Filters, Sterile
    Replacement Filters, Sterile
    76193-124
    4-000-051-S
    Pipet-Aid
    Log in for availability
    Log in for availability
    Paquete de 5
    Paquete de 5 - $1,533.48
    $1,533.48
    Paquete de 5
       $1,533.48
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Replacement Filters, Nonsterile
    Replacement Filters, Nonsterile
    53507-080
    4-000-052
    Pipet-Aid
    4-000-052
    Log in for availability
    Log in for availability
    Paquete de 50
    Paquete de 50 - $7,587.95
    $7,587.95
    Paquete de 50
       $7,587.95
    Specifications:
    Accessories