Clamps, [ST], Metal, Ace Glass Incorporated
Proveedor: Ace Glass
7599-25 7599-13
89059-624PK 1862.17 MXN
89059-624 89059-622
Clamps, [ST], Metal, Ace Glass Incorporated
Abrazaderas Pinzas para juntas NS
A nickel plated steel clip that snaps on and off standard taper joints with ease and offers increased resistance to heating and cracking
Can be used to 200°C. Will not scratch glass.