Cylindrical Reaction Flask,Duran Flange, Ace Glass Incorporated
Proveedor: Ace Glass
6521-14 6521-35 6521-20 6521-30 6521-25 6521-10 6521-27 6521-16 6521-12
89055-128EA 31277.49 MXN
89055-126 89055-136 89055-124 89055-134 89055-122 89055-132 89055-130 89083-520 89055-128
Cylindrical Reaction Flask,Duran Flange, Ace Glass Incorporated
Recipientes de reacción Vasos de Reacción Cilíndricos
Rugged cylindrical reaction flasks feature Duran style ground flange with O-Ring groove, designed for use with a quick release clamp
Flask sizes 10L and larger are supplied with capacity reference graduations. Flask sizes 30L and larger have a reduced area to flange. All vessels feature straight sides and are manufactured from borosilicate glass.
Información para pedidos: The 1L, 2L, and 3L flasks are designed for use with Clamp 89055-110. All larger flasks are designed for use with Clamp 89055-114. The 1L, 2L, and 3L flasks are supplied with CAPFE O-Ring 89061-166. All larger flasks are supplied with CAPFE O-Ring 89061-172.