A-1 Medium Broth MPN Tubes, Hach
Proveedor: REACTIVOS Y EQUIP SA DE CV MX
2560915 2560915
97026-334EA 33.73 MXN
97026-334 CA97026-334
A-1 Medium Broth MPN Tubes, Hach
Medios Medios preparados
A-1 Medium broth tubes provide detection of Fecal Coliform Bacteria using the Most Probable Number (MPN) method.
Save time and labor costs with ready-to-use MPN tubes, with minimal training requirements.