Flask, Dewar, Ace Glass Incorporated
Proveedor: Ace Glass
7075-10 7075-20 7075-05 7075-25 7075-15
89057-552EA 21747.05 MXN
89057-552 89057-550 89057-546 89057-544 89057-548
Flask, Dewar, Ace Glass Incorporated
Dewars y recipientes criogénicos
Dewar flask
Cylindrical form borosilicate glass is silvered and evacuated. Furnished with an aluminum base and plastic mesh. Intended for use with liquid nitrogen only.