CiDecon® Concentrated Disinfectant, Decon Labs
8504
89233-188CS 6500.14 MXN
89233-188
CiDecon® Concentrated Disinfectant, Decon Labs
CiDecon® Concentrated Phenolic Disinfectant
Concentrated germicidal cleaner for disinfecting surfaces in lab production areas and on process equipment. Diluted one ounce per gallon of water (1:128), CiDecon® is effective against a wide range of pathogenic organisms including TB, HIV-1, bacteria, viruses, and fungi. Conforms to OSHA’s Bloodborne Pathogens Standard. EPA registered.