NEUTRAD® Neutral pH Liquid Cleaner, Decon Labs
Proveedor: Decon Labs
NEUTRAD®
3002 3001 3003 3001 3003 3002
89234-808CS 5793.18 MXN
CA89234-806 89234-808 CA89234-808 89234-804 CA89234-804 89234-806
NEUTRAD® Neutral pH Liquid Cleaner, Decon Labs
Soluciones de limpieza Detergentes y Limpiadores de Superficie
NEUTRAD® is a safe detergent for manual scrub cleaning of any labware – replaces old fashioned caustic soap powders! Highly concentrated, very economical. Near-neutral, pH is skin-safe! Phosphate-free, very rinsable and biodegradable. Liquid NEUTRAD® is instantly soluble, totally rinsable, safe, near-neutral pH and phosphate-free. Made with recycled packaging.