Impinger, Greenburg-Smith, Ace Glass Incorporated
Proveedor: Ace Glass
7536-31 7536-15 7536-13 7536-16 7536-06 7536-26 7536-25 7536-10 7536-04 7536-12 7536-32
89059-342EA 3192.6 MXN
89059-348 89059-358 89059-346 89059-356 89059-354 89059-342 89059-352 89057-710 89059-362 89059-350 89059-360
Impinger, Greenburg-Smith, Ace Glass Incorporated
Burbujeadores
A high velocity impinger used to determine the dust concentration in air, or in other gases
A glass plate is located at a distance of 5 mm below the orifice tip so that incoming particles are impinged onto the plate and are momentarily arrested, after which they are washed into the liquid. Total volume is approximately 500 mL. Bottle has a reinforced [ST] 45/50 outer joint at top. Inlet and outlet are both 12.5 mm O.D. Fabricated from borosilicate glass.
Información para pedidos: "Standard" version has straight tubes on inlet and outlet. "Modified" versions have [SJ] ball or [SJ] socket. Tube is available with impinging plate, or without impinging plate (cut flat). Available as a complete item with bottle and tube, or components can be purchased separately.