Drawer/Ganizer, Clamp / Instrument Tray, Spectrum Laboratories
155525 155600
89219-752EA 1805.59 MXN
89219-740 89219-752
Drawer/Ganizer, Clamp / Instrument Tray, Spectrum Laboratories
Spectrum introduces new Drawer/Ganizer Portable Trays molded from thicker 1/8 inch heavy duty polystyrene with easy-gripping folded edges for transporting assorted lab items and mobilizing ex- perimental activities
Two spacer sizes available, each with 3 compartment sizes for storing clamps, hardware, instruments and other odd-shaped items.
The deeper compartments are better suited than most other lab trays for holding larger quantities as well as serving as secondary containment for spills. Like the original Drawer/Ganizer Spacers, these Portable Organizer Trays are designed for standard lab drawers, refrigerators and bench tops to organize and protect supplies, conserve valuable space and promote lab safety. Spectrum offers 6 configurations in two sizes (1 tray/pkg).