Drawer/Ganizer, Parts / Chromatography Tray, Spectrum Laboratories
Proveedor: Spectrum
155645
89219-744
Drawer/Ganizer, Parts / Chromatography Tray, Spectrum Laboratories
Bandejas/Platos/Ollas Bandejas para organización de cajones
Spectrum introduces new Drawer/Ganizer Portable Trays molded from thicker 1/8 inch heavy duty polystyrene with easy-gripping folded edges for transporting assorted lab items and mobilizing ex- perimental activities
Various size compartments for easy to find storage of lab & chromatography supplies like utensils, valves, connectors, etc.
The deeper compartments are better suited than most other lab trays for holding larger quantities as well as serving as secondary containment for spills. Like the original Drawer/Ganizer Spacers, these Portable Organizer Trays are designed for standard lab drawers, refrigerators and bench tops to organize and protect supplies, conserve valuable space and promote lab safety. Spectrum offers 6 configurations in two sizes (1 tray/pkg).