Drawer/Ganizer, Micropipettor Tray, Spectrum Laboratories
155675
89219-746
Drawer/Ganizer, Micropipettor Tray, Spectrum Laboratories
Spectrum introduces new Drawer/Ganizer Portable Trays molded from thicker 1/8 inch heavy duty polystyrene with easy-gripping folded edges for transporting assorted lab items and mobilizing ex- perimental activities
2 sizes designed for storing up to 7 micropipettors, pipettes and/or glass syringes as well as 2 types of pipette tips.
The deeper compartments are better suited than most other lab trays for holding larger quantities as well as serving as secondary containment for spills. Like the original Drawer/Ganizer Spacers, these Portable Organizer Trays are designed for standard lab drawers, refrigerators and bench tops to organize and protect supplies, conserve valuable space and promote lab safety. Spectrum offers 6 configurations in two sizes (1 tray/pkg).