O-Rings, FETFE®, Ace Glass Incorporated
Proveedor: Ace Glass
7855-715 7855-702 7855-772 7855-746 7855-742 7855-740 7855-748 7855-704 7855-744 7855-738 7855-764 7855-774 7855-734 7855-705 7855-716 7855-750 7855-712 7855-739 7855-787 7855-727 7855-708 7855-722 7855-720 7855-730 7855-721 7855-709 7855-718 7855-726 7855-714 7855-707 7855-706 7855-775 7855-766 7855-782 7855-771 7855-710 7855-701 7855-703 7855-747 7855-711 7855-732
89060-976PK 580.72 MXN
89061-028 89061-026 89061-004 89061-048 89060-980 89060-982 89060-960 89060-966 89060-988 89060-968 89061-060 89060-984 89060-962 89060-964 89061-020 89061-042 89061-062 89061-046 89061-002 89061-022 89061-038 89061-016 89061-036 89061-014 89060-954 89060-976 89060-998 89060-956 89060-994 89061-070 89060-972 89060-996 89060-952 89060-974 89061-030 89061-074 89061-056 89061-034 89060-958 89061-012 89061-054
O-Rings, FETFE®, Ace Glass Incorporated
Juntas Anillos de Sellado
FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. Generally black or white in color.