Separator Depth Filters, Polypropylene/Polyethylene, Cytiva (Formerly Pall Lab)
Proveedor: Cytiva
60344 60343 60344 60343
97015-514EA 93.81 MXN
CA97015-514 97015-514 CA97015-512 97015-512
Separator Depth Filters, Polypropylene/Polyethylene, Cytiva (Formerly Pall Lab)
Filtros Filtros de membranas
The Polypropylene/Polyethylene Separator Depth Filters have outstanding wet strength and can be folded or handled the same as laboratory paper. Because of their material of construction, these disc filters are virtually inert to chemical and biological agents.
  • Outstanding wet strength
  • High chemical resistance
  • Channeling effect
Their high chemical resistance and wet strength ensure stability when filtering concentrated acids, alkalis, or oxidizers.
When used as prefilters in series with submicron membrane filters, the PP/PE depth filters increase the overall filter flow rate by providing a channeling effect for particulate-laden and viscous fluids.
These filters can also be used as separators between stacked discs in a single filter holder during serial filtration. Maximum temperature value represent the value of maximum operating temperature.
Filter media is a blend of Polypropylene and Polyethylene.