Mantener la sesión abierta?
Finalizando en 
Su sesión ha finalizado
Su sesión ha caducado. Por seguridad lo hemos desconectado.
¿Quiere conectarse de nuevo?
VWR® 96-Well Polypropylene Plates
Proveedor: VWR International
V07-6000 V07-1100 V07-0067N V07-5000 V07-0063N V07-0011N V07-0017N V07-0060N V07-3900 V07-0066N V07-0065N V07-1000 V07-7400 V07-1200 V07-2000 V07-0010N V07-8400 V07-0061N
89237-514PK 1718.33 MXN InStock
89237-518 89237-506 89237-548 89237-516 89237-542 89237-530 89237-534 89237-538 89237-514 89237-546 89237-544 89237-504 89237-522 89237-508 89237-510 89237-528 89237-526 89237-540
VWR® 96-Well Polypropylene Plates
Microplacas Placas Deep Well
VWR® 96-Well Polypropylene Plates are manufactured in a variety of sizes and well formats to meet the demanding requirements of modern laboratory applications. VWR® Covers for 96-Well Plates are offered in either Ethylene Vinyl Acetate or Silicone with a sprayed-on Teflon barrier.
  • All covers fit interchangeably
  • Round, conical, and flat bottom formats are available
  • Virgin polypropylene microplate construction
  • Covers are available in multiple materials and thicknesses
These virgin polypropylene microplates offer a variety of sample volumes that are uniform dimensionally so that all covers, whether round or square, fit interchangeably on other round or square well microplates. This reduces the number of covers needed for most laboratories. There are shallow well microplates with flat, round, and conical bottoms for volumes up to 350µL. The medium well microplate has a round bottom for volumes up to 500µL. The deep well microplates are available in round well with round bottoms for volumes up to 1.0mL and square well with either a conical or round bottom for volumes up to 2.0mL.
The EVA microplate covers are designed to stabilize samples during transport from one workstation to another with the option of a thicker cover for seal strength that is not piercible or a thinner version that is piercible by most autosampler needles. The Silicone covers are available in two thicknesses, both of which are available either pre-cut or non-cut for resealability. All round well and square well covers fit all round well and square well microplates in the VWR Collection. All items are sold in packs of ten covers.
Información para pedidos: All microplates, and all covers, are sold in packs of ten.
Order Now
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
PEDIDO
INFORMACIÓN RELACIONADA
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
O
Revise y compare las alternativas
Disponemos de alternativas con las que puede ahorrar hasta  por producto unitario.  Para comparar detalles de los productos, seleccione hasta 3 alternativas y haga clic en comparar seleccionados. Para añadir productos a su cesta, introduzca la cantidad y haga clic en Añadir a la cesta.
Producto Original:
Descripción Numero del catalogo Disponibilidad Unidad Your Price
(IVA no incluido)
Precio Por Uds.