UVP Portable UV Lamps, Analytik Jena
Proveedor: Analytik Jena
95-0009-02 95-0010-02 95-0011-01
36585-518EA 13107.86 MXN
36585-518 36585-516 36585-267
UVP Portable UV Lamps, Analytik Jena
Luces Lámparas de rayos UV
Portable UV Lamps use one ultraviolet tube: midrange, longwave or shortwave
  • Dimensions: 24.1L x 7.1W x 23.8H cm (91/2 x 213/16 x 93/8")
An optional charger (12 V float battery charger) is available for charging rechargeable type batteries.
The lamps are designed with a distinctive wrap-around handle which comfortably balances the lamp in your hand. These lamps operate by two 6-Volt standard or rechargeable batteries (batteries ordered separately). A built-in flashlight is included. A sliding wavelength selector is attached to UVLG-48 multi-band model for switching between longwave and shortwave UV.