Ubbelohde ASTM Series Capillary Viscometers, Fungilab
Proveedor: Fungilab
CV003-008 CV003-013 CV003-010 CV003-007 CV003-015 CV003-006 CV003-016 CV003-011 CV003-009
89236-832EA 7205.96 MXN
89236-850 89236-840 89236-830 89236-832 89236-844 89236-834 89236-836 89236-848 89236-838
Ubbelohde ASTM Series Capillary Viscometers, Fungilab
Viscosímetros Viscosímetros capilares
Ubbelohde viscometers are used to determine kinematic viscosities of transparent Newtonian liquids such as jet and hydraulic lubricants.
Certificaciones: UNE 400313, ASTM D445-446, ASTM D2515 and ISO 3104-3105).
Información para pedidos: Includes constant value calibration certificate at 40ºC (104ºF) and 100ºC (212ºF).