U-Tube Viscometer Type BS/U Capillary Viscometers, Fungilab
Proveedor: Fungilab
CV006-103 CV006-105 CV006-106
89236-960EA 5382 MXN
89236-960 89236-962 89236-956
U-Tube Viscometer Type BS/U Capillary Viscometers, Fungilab
Viscosímetros Viscosímetros capilares
BS U Tube Reverse Flow capillary viscosmeter for opaque liquids
Made of borosilicate glass, and non-alterable timing mark.
Certificaciones: BS, UNE 400313, ASTM D-445, D446, ISO 3104, ISO 3105.
Información para pedidos: Includes constant value calibration certificate at 40ºC (104ºF) and 100ºC (212ºF).