Flexible Motor Drive Shaft, Ace Glass Incorporated
Proveedor: Ace Glass
8081-12 8081-08 8081-15 8081-27 8081-30 8081-07 8081-06 8081-24 8081-05 8081-32 8081-19
89062-048EA 911.14 MXN
89062-036 89062-046 89062-038 89062-048 89062-050 89062-052 89062-040 89062-032 89062-042 89062-034 89062-044
Flexible Motor Drive Shaft, Ace Glass Incorporated
Imánes de agitación Ejes de agitación
Fully flexible drive shaft connects your stirring motor to any size or type of reactor stir shaft
The shafts operate up to 14,000 rpm, and they should be run in a counter-clockwise direction. Typical torque ratings — sharp bend in shaft, (4-inch loop): 4.7 Kg-cm, (4 in-lbs); Straight shaft: 28 Kg-cm, (24 in-lbs). Shafts measure approximately 91.4 cm, (36 inches) or 152.4 cm, (60 inches) with hand-piece and motor coupling attached.
Designed with a ball bearing motor coupling at one end for connection to any motor with an 8mm, (5/16-inch) diameter shaft. The other end has a detachable hand-piece with an 8mm round steel pin adapter for connection to chucks, (must be purchased separately). The hand-piece can be supported by a standard lab clamp, or hand held.
Información para pedidos: Complete units consist of either 91.4 cm shaft (89062-032) or 152.4 cm shaft (89062-034), motor coupling for 8mm motor shaft (89062-040), hand-piece with 1/4-inch collett and adapter (89062-038), chuck wrench with chained key (89062-042). Operating and lubrication instructions are also included. Optional adapter 89062-048 allows for connection to motors with 9.5mm (3/8-inch) O. D. shaft. All items may be purchased separately.