Analysis packages are ideal solutions for getting top results, and are designed for a wide variety of tests to fit the unique needs of your samples.
- Simple to operate and maintain
- Consistently accurate and reliable measurements
- Configurable to each user’s unique needs
- VWR® three-year limited warranty
Low Ionic Strength / Ultrapure Sample Package: All you need to get results that are up to your standards.
The VWR® sympHony® Low Ionic Strength / Ultrapure Sample Package is designed to fit the unique needs of testing pure samples. This package includes a refillable Ag/AgCl reference glass body pH electrode with integrated temperature sensor. The sleeve diaphragm of the electrode allows for high flow of the reference solution, mitigating drift, slow response times, non-reproducible and inaccurate readings that are common in low ionic strength measurements. Extreme pH Package: A complete solution for any pH range. The VWR® sympHony® Extreme pH Package is ideally suited to perform regardless of sample pH. This package includes a refillable Ag/AgCl reference glass body pH electrode with integrated temperature sensor, designed to minimize the effects of acid and alkaline errors.
Difficult Sample Package: A complete solution for a wide variety of sample matrices. The VWR® sympHony® Difficult Sample Package is designed to work well in sample matrices such as biological buffers and enzymes, viscous or non-aqueous samples, and samples containing sulfides. This package includes a refillable Ag/AgCl reference glass body pH electrode with integrated temperature sensor. The sleeve diaphragm of the electrode allows for high flow of the reference solution while the silver ion barrier prevents precipitation of the filling solution when used in these difficult applications.
General-Purpose Package: A simple, easy to use all purpose bundle. The VWR® sympHony® general-purpose Package is designed to meet the needs of most common pH applications. The package includes a low-maintenance epoxy body non-refillable electrode with integrated temperature sensor.
High Throughput Package: Simplified measurement for high-throughput labs. The VWR® sympHony® high-throughput Package is designed for taking multiple measurements including pH, conductivity, ISE, or ORP.
Portable pH Package: For all your on-the-go pH testing needs. The VWR® sympHony® Portable pH Package is designed for testing pH in the field. This package comes with a low maintenance non-refillable polycarbonate body electrode with integrated temperature sensor.
Certificaciones: VWR® sympHony benchtop meters meet IP 54 specifications for protection from dust and water while handheld meters meet IP 67 specifications. Both are CE and cETLus marked.