Mantle, Aluminum Housing, Solid Bottom, Ace Glass Incorporated
Proveedor: Ace Glass
12053-23 12053-25 12053-13 12053-17 12053-19 12053-26 12053-21 12053-05 12053-07
89075-958EA 26148.68 MXN
89076-030 89084-348 89075-950 89076-028 89075-944 89076-026 89075-946 89075-948 89075-958
Mantle, Aluminum Housing, Solid Bottom, Ace Glass Incorporated
Calentadores Capas calefactoras
Solid, aluminum-housed heating mantles are versatile enough to be used on lab benches, stands, or floors
They accommodate cylindrical and spherical flasks. Mantles must be operated through a temperature controller, and offer an operating temperature range of 10° to 450°C. All units are supplied with a 122 cm (4') detachable cord with interlocking connector.
Certificaciones: CSA approved.