Trubore® Bearing, with Tooled Lubricating Cup Top and Jointed Bottom, Ace Glass Incorporated
Proveedor: Ace Glass
8039-05 8039-25 8039-10 8039-35 8039-03 8039-20
89061-712EA 2915.97 MXN
89061-698 89061-712 89061-700 89061-718 89061-694 89061-708
Trubore® Bearing, with Tooled Lubricating Cup Top and Jointed Bottom, Ace Glass Incorporated
Imánes de agitación Herramientas de agitación
Interchangeable ground joint bearing for use with precision ground stirring shafts
  • Recommended top stirring speed, with appropriate stirrer lubrication is 1500 rpm
  • Manufactured from borosilicate glass
Bearings feature a tooled lubricating cup at the top and a joint at bottom.