VWR® Contour Polypropylene Swivel Chairs
Proveedor: VWR
VWR® Contour™
VSGG-H-R
89201-121EA 17244.09 MXN
89201-121
VWR® Contour Polypropylene Swivel Chairs
Sillas
This 100% polypropylene chair has anti-microbial properties and UV inhibitors.
  • Lumbar Support Included
  • Waterfall Front
  • Seat Width: 17½"
  • Seat Diameter: 15½"
It features a 26” diameter composite base with casters and a pneumatic seat height adjustment.
Medium and high-bench-height stools have 50.8 cm, 20" diameter footrings with adjustable heights.
Información para pedidos: Contour seating is covered by a five-year warranty against defects in material, workmanship or abnormal wear. Upholstery is covered for one year. Pneumatic cylinders are covered for the life of the product.