Protector® Fume Hood Airflow Monitors, Service Fixture Kits, and Duplex Receptacle Kits, Labconco
Proveedor: Labconco
9809200 9413300 9808400 9808700 9851100 9808500 9827900 9410300 9828000 9851500 9413400 985340010814 985770010814 985420010814 9809700 9808800 9808300 9809300 9808900 9809100
89260-114EA 3284.06 MXN
89260-114 89260-786 89260-104 89260-112 89260-102 89260-118 89260-108 89260-116 89260-106 89260-092 89260-090 89260-084 89260-096 89260-082 89260-094 89260-088 89260-110 89260-100 89260-086 89260-098
Protector® Fume Hood Airflow Monitors, Service Fixture Kits, and Duplex Receptacle Kits, Labconco
Vitrinas de gases y recintos Campanas de gases químicos
There are many accessories available for customizing Protector® Laboratory Hoods including Service Fixture Kits, Duplex Receptacle Kits and Guardian™ Airflow Monitors.
Each Service Fixture Kit includes one remotely-controlled service fixture or gooseneck with valve and outlet tubing, color-coded fixture knob and color-coded hose connector. Choose from a variety of configurations. All kits are compatible with Protector® Premier®, XStream®, PVC and XL™ Hoods. Gooseneck kits are also compatible with Protector® Stainless Steel Perchloric Acid Hoods.
Duplex Receptacle Kits include wiring. Guardian™ Airflow Monitors alert operators to low airflow conditions. Guardian™ Digital Airflow Monitors provide a digital display of face velocity as well as an audible alarm.
Información para pedidos: Base cabinets, blowers and work surfaces are sold separately.