REDISHIP Protector® Premier® Laboratory Hoods and REDISHIP SpillStopper™ Work Surfaces, Labconco®
Proveedor: Labconco
10050000010814 10060000010814 10080000010814 10040000010814
89260-052EA 254409.16 MXN
89260-052 89260-050 89260-056 89260-054
REDISHIP Protector® Premier® Laboratory Hoods and REDISHIP SpillStopper™ Work Surfaces, Labconco®
Vitrinas de gases y recintos Campanas de gases químicos
Popular models of Protector® Premier® Laboratory Hoods and supporting matching work surfaces (sold separately) are available from VWR® stock for immediate shipment
REDISHIP SpillStopper™ Work Surfaces are dished to contain spills and feature a left rear corner cutout and oval cup sink with cover.
REDISHIP Protector® Premier® Fiberglass Hoods feature left and right-sided GFCI duplexes and Labconco's signature one-piece molded fiberglass reinforced polyester liner that provides superior corrosion and chemical resistance and durability. Each hood is factory prepared to accommodate up to eight service fixtures (sold separately). The hood's by-pass block design restricts airflow, allowing the REDISHIP Protector Premier to operate as a reduced air volume (RAV) or variable air volume (VAV) system with no modifications.
Información para pedidos: Base cabinets are sold separately. Choose from service fixture kits and other accessories for customization (sold separately). Fiberglass and Coated Steel Blowers in sizes matched to most installation requirements are also available (sold separately).