Moticam® 2 Digital Microscope Camera, Motic Instruments
Proveedor: Motic
Moticam®
1100600100611
89238-598EA 11086.92 MXN
89238-598
Moticam® 2 Digital Microscope Camera, Motic Instruments
Cámaras
Moticam® attachable cameras allow the user to turn almost any existing micrscope into a digital microscope
The Moticam 2 is a 2MP camera for those who are looking for a large display and crisp imaging sensor. It has a 1/3” CMOS sensor, a live resolution of 2MP, a 12mm focusable lens, and is compatible with Windows XP and higher and also with Mac OSX.
These solutions contain everything necessary to get started, including our powerful imaging software. Connect to the eyepeice, a trinoculor mount or use the camera on its own with a gooseneck as a Macro-Camera
Certificaciones: UL Listed.
Información para pedidos: Camera is supplied with all accessories listed above. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a 1 year electrical and 5 year mechanical warranty.