Vacuum Trap, Dewar Type, Ace Glass
Proveedor: Ace Glass
8757-10
89063-974EA 8698.56 MXN
89063-974
Vacuum Trap, Dewar Type, Ace Glass
Trampas de vacío
This Dewar-type vacuum trap has a [ST] 24/40 joint at the bottom
The inside diameter of the coolant section is 50mm, and length is approximately 250mm. The O.D. of the inlet and outlet is 22mm. Fabricated from borosilicate glass.